Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® pHenomenal® OX 4100L Dissolved Oxygen Meter, Bench

Supplier: Avantor
VWR® pHenomenal®
1BV300Y
76460-490EA 1657.83 USD InStock
76460-490
VWR® pHenomenal® OX 4100L Dissolved Oxygen Meter, Bench
Electrochemical Meters Oxygen Meters
This user friendly instrument with articulated electrode stand and IP 43-rated housing provides high resolution and accuracy for precise measurements. With USB output for connection to a computer. GLP compliant.

  • Large LCD graphic display with continuous LED backlight showing both DO and temperature values simultaneously
  • Data logging function selectable between 1 min and 1 h
  • Memory capacity of 5000 data sets
  • Automatic power shut off: Adjustable from 10 min to 24 h
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when re-calibration is required
  • Low energy consumption. Choice of mains supply or 4× 'AA' batteries for over 1000 operating hours

Delivery information: Ready to use oxygen OX 4100 L kit (76460-490) includes meter, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 30 ml cleaning solution, 50 ml electrolyte, 3 exchange caps, plugin power supply, DO electrode OXY 11.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty