Would you like to log in again?
DISCHARGE UNIT PROSPENSR PLUS 3MM TUBING
Supplier: SARTORIUS CORPORATION EDI
Discharge unit/Dispenser Head, Prospenser Plus, 3.0mm Spiral Tubing for FEP (Fluoroethylene propylene) compatible substances. Suitable for all Bottle-top Dispensers models.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DISCHARGE UNIT PROSPENSR PLUS 3MM TUBING 76460-540 LH-721649 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty