Would you like to log in again?
EXTERNAL THREAD CAP NAT NONST 2ML CS6000
Supplier: Greiner Bio-One
External Thread Cap, New Decapper Compatible, Natural, Non-Sterile, Suitable for storage at -20 deg C, -80 deg C and in the vapour phase of LN2, Ultra-pure medical grade, USP class VI certified polypropylene w/o leachables, Free of detectable human DNA, endotoxins, DNase, CE marked, Size: 2 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EXTERNAL THREAD CAP NAT NONST 2ML CS6000 76460-878 305378-923 Case of 6,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty