Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ultra-Duty GHS Chemical Waterproof and UV Resistant Labels, Essendant
Supplier: Essendant
Avery®
AVE60517
76460-880PK 77.97 USD InStock
76460-880
Ultra-Duty GHS Chemical Waterproof and UV Resistant Labels, Essendant
Labels
Waterproof and resistant to chemicals, abrasion, sun, tearing, and harsh temperatures.
  • Ideal for labeling drums, totes, pails, jugs, and bottles
  • High-performance adhesive
Easy Peel® design for fast peeling and convenient use with laser printers. Rectangular labels are supplied on letter-sized, 8.5×11" sheets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty