Would you like to log in again?
Repositionable Vinyl Cloth Labels, 3" Core, Brady
Supplier: Brady
THT-87-498-5 THT-91-498-5 THT-1-498-10 THT-68-498-10 THT-90-498-5 THT-165-498-5
76461-100RL 384.44 USD InStock
76461-100 76461-166 76461-102 76461-096 76461-098 76461-094
Repositionable Vinyl Cloth Labels, 3" Core, Brady
Labels
Permanent or repositionable once applied, B-498 CleanLift® Series Vinyl Cloth remains securely in place until repositioning is required.
  • Repositionable
The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning. Sticks to any clean, dry surface. Specially formulated topcoat for excellent print quality. Semi-gloss finish for crisp, clean look.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 76247-389 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use CA11026-100 for a different printer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty