Would you like to log in again?
VWR® EZLabpure™ Round Carboys, with 83B VersaCap®
Supplier: Avantor
VWR®
155-C117-FLS 155-C817-FLS 155-B117-FLS 155-E191-FLS 155-B817-FLS 152-C191-FLS 152-B191-FLS
76461-398EA 84.86 USD InStock
76461-398 76461-400 77296-504 77296-502 77296-500 76461-402 76461-404
VWR® EZLabpure™ Round Carboys, with 83B VersaCap®
Bottles Carboys
VWR® EZLabpure™ round carboys are manufactured using high-purity virgin USP Class VI medical grade polypropylene.
  • Carboys can be used for storing and shipping reagents, chemicals, culture media, and standards
  • Polypropylene carboys are autoclavable at 121 °C for twenty minutes
  • Can be used at a temperature below 0 °C
  • Superior chemical resistance
A premium line of high-quality, round HDPE and polypropylene (PP) laboratory carboys with the patented VersaCap® technology. These durable, rugged, and shatterproof round carboys are a great alternative to glass carboys. They can be used for mixing, storing, and dispensing liquids in volumes from 10 to 50 liters. These carboys are available with and without a spigot.
Certifications: UPS Class VI materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty