Would you like to log in again?
VWR CARBOY PP W/WHITE CAP WIDE MOUTH 20L
Supplier: VWR International
Carboy, with White Cap, Wide Mouth, Without Spigot, Material: Polypropylene, premium line of high quality, Round Wide Mouth Polypropylene (PP) Laboratory Carboys. Our versatile carboys with strong easy-grip handles aim to take full advantage of storage, allow for easy efficiency, Size: 20L
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CARBOY PP W/WHITE CAP WIDE MOUTH 20L 76461-408 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty