Would you like to log in again?
VWR BAG AUTCLV CLEAR 2MIL PP 12X24 CS500
Supplier: VWR International
Autoclave bag, PreSterilized (gamma irradiated process), designed for Clean Room applications. Clear 2 mil (51UM) polypropylene (plain clear bags) for processing at 275 deg f (235 deg C) in autoclave. Packaged in 10 sealed sleeves of 50 bags with GIP indicator, 12x24in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR BAG AUTCLV CLEAR 2MIL PP 12X24 CS500 76461-648 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty