Would you like to log in again?
VWR BAG AUTCLV RD BIO 3MIL PP 37X48 CS50
Supplier: VWR International
Autoclave bag, PreSterilized (gamma irradiated process), designed for Clean Room use. Printed BIOhazard bags meet all Federal, State and territorial regulations. Red 3 mil (51UM) polypropylene for autoclave processing at 275 f (235 C). Packaged in 1 sealed sleeves of 50 bags with indicator, 37x48in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR BAG AUTCLV RD BIO 3MIL PP 37X48 CS50 76461-664 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty