Would you like to log in again?
HardyCHROM™ Bcc, Chromogenic Medium for Burkholderia cepacia, Hardy Diagnostics
Supplier: Hardy Diagnostics
G335
76461-868PK 56.52 USD InStock
76461-868
HardyCHROM™ Bcc, Chromogenic Medium for Burkholderia cepacia, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
HardyCHROM™ Bcc is a chromogenic agar for selective isolation and differentiation of Burkholderia cepacia and 28 other Bcc strains, including B. cenocepacia and B. multivorans.
  • Easy detection of pink-to-magenta colonies
  • Useful for testing non-sterile, water based pharmaceuticals and cosmetics
  • Inhibitory for the growth of gram-positive organisms
USP-60 describes tests to aid in detecting members of the Bcc group in non sterile products.
Caution: This media is to be stored and incubated in the dark. If you have a light in your refrigerator, incubator, or you leave the media exposed to light on the bench top, Hardy Diagnostics offers free Blok-Boxes upon request. Blok-Box is a reusable cardboard box that holds one sleeve of ten Petri plates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty