Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acid-Based Cleaner and Disinfectant, CIP 200™, STERIS®

Supplier: Steris

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CIP 200®
1D2302 1D23D3 1D2305 1D2308 1D2301
76461-892EA 5537.59 USD InStock
76461-892 76461-894 76461-890 76461-886 76461-888
Acid-Based Cleaner and Disinfectant, CIP 200™, STERIS®
Cleaning Agents Detergents and Surface Cleansers
CIP 200® is a high performance, phosphoric acid cleaner for use in manual, immersion, and recirculating spray equipment including clean-in-place (CIP) systems.

  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues CIP, clean-out-of-place (COP), and manual applications
  • Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Global Availability - and manufactured with biodegradable surfactants to support harmonization of cleaning processes

The blended acid and surfactant system rapidly and efficiently removes a wide variety of scales, particulate, and inorganic soils typically found in pharmaceutical, biotechnology, cosmetic, and food and beverage processes.

Certifications: USDA approved under Category A3. Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility. EPA Biocidal and virucidal claims on washable, hard, nonporous surfaces in the presence of 5% organic soil (serum) when used at a 1% concentration of product for the following:
- Staphylococcus aureus (ATCC 6538) in 10 minutes at or above 60 °C
- Pseudomonas aeruginosa (ATCC 15442) in 10 minutes at or above 60 °C
Virucidal claim
- Bovine Viral Diarrhea Virus (BVDV) at 10 minutes at or above 60 °C

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty