Would you like to log in again?
AssureWear™ SMS Isolation Gowns, Medicom
Supplier: Medicom
A69972
76461-928CS 324.16 USD InStock
76461-928
AssureWear™ SMS Isolation Gowns, Medicom
Disposable Garments
Provides reliable protection without compromising comfort.
  • Single-use gown
  • AAMI Level 2
  • Features neck ties, elastic cuffs, and full back closure
  • Sewn seams ensure superior strength
  • Latex-free
These 3-layer SMS Isolation gowns feature a tape tab neck, elastic cuffs, and full back. Single-use gowns are designed to provide reliable protection without compromising comfort. Sewn seams ensure superior strength.
Disposable SMMS Polypropylene Isolation Gown designed to protect health professionals and patients from cross infection.
Packaging: Gowns are packaged 10/bag, 10 bags/case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty