Would you like to log in again?
Tube Roller-Mixer, Globe Scientific
Supplier: Globe Scientific
GTRM-FS
76462-034EA 281.91 USD InStock
76462-034
Tube Roller-Mixer, Globe Scientific
Shakers and Mixers Rotating/Rocking Shakers
The GTRM-FS Roller-Mixer combines gentle rocking with continuous rolling motion to ensure your samples are thoroughly mixed for optimal homogeneity.
  • Warranty: 2 Years
The unique design of the Globe Scientific GTRM-FS rollermixer combines the operation of a vacuum tube roller with a see-saw rocking platform to allow for the ultimate in homogeneity of your blood/urine tubes and other sample types. This combined motion also helps ensure higher culture yields. The GTRM-FS is compatible for use in cold rooms and non-CO₂ incubators. The rolling motion is a fixed 36 rpm and the rocking motion is set at ±11°.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty