Would you like to log in again?
HyClone™ SFM4HEK293, HEK 293 Media, Cytiva
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyClone™
SH30521.02 SH30522.01 SH30522.01 SH30521.02
82003-356EA 115.84 USD InStock
CA82003-356 82003-356 CA76462-526 76462-526
HyClone™ SFM4HEK293, HEK 293 Media, Cytiva
Cell Culture Media
HyClone™ SFM4HEK293 is a protein-free cell culture media without animal-derived components (ADCF) used to support production of adenoviral vectors and recombinant proteins in HEK 293 cultures.
  • Developed through metabolic pathway design process for high adenoviral vector and protein production
  • Ready-to-use liquid formulation requires no supplementation
  • Allows for direct or sequential adaptation
  • For large-scale culture applications, including perfusion and fed-batch strategies
  • Manufactured according to cGMP guidelines
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty