Would you like to log in again?
HyClone™ CDM4HEK293, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH30859.01 SH30858.02 SH30858.02 SH30859.01
95026-630EA 119.95 USD InStock
CA95026-630 95026-630 CA76462-536 76462-536
HyClone™ CDM4HEK293, HyClone products (Cytiva)
Cell Culture Media
HyClone™ CDM4HEK293 media are chemically-defined, animal-derived component free (ADCF) cell culture media.
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
  • Metabolically designed for high cell yield and recombinant protein/vector production
HyClone CDM4HEK293 is a regulatory-friendly medium developed through the Metabolic Pathway Design to support the extremely high productivity and cell density in suspension cultures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty