Would you like to log in again?
HyClone™ HyCell TransFx-H Medium, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH30944.01 SH30944.01 SH30939.02 SH30939.02
10842-656EA 125.74 USD InStock
10842-656 CA10842-656 76462-546 CA76462-546
HyClone™ HyCell TransFx-H Medium, HyClone products (Cytiva)
Cell Culture Media
HyClone™ HyCell™ TransFx-H media supports transient transfection, growth, and expression of a variety of recombinant proteins in HEK 293 cells. It is a high-performing, animal-derived component-free (ADCF) cell culture media with the versitility to be used with a variety of HEK 293 cell lines in microlitre volumes starting from 200 µl to production-scale volumes in bioreactors.
  • Defined hydrolysate-free ADCF cell culture media formulation
  • Supports transient transfection and growth of multiple HEK 293 cell lines
  • Supports the transient expression of a variety of recombinant proteins
  • Same media for transient transfection and production
  • Developed through our Metabolic Pathway Design process for high transfection efficiency, high viable cell yields, and recombinant protein production
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
HyCell TransFx-H media is a hydrolysate-free and regulatory-friendly Cytiva ADCF cell culture media. Its versatility allows quick adaptation and supports excellent transfection, high viable cell density, and high productivity across a variety of HEK293 cell lines.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty