Would you like to log in again?
HyClone™ Cell Boost™ 7a and 7b Feed Supplements, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyClone™
SH31026.01 SH31027.07 SH31026.01 SH31026.07 SH31026.07 SH31027.07 SH31027.04
76462-552EA 1366.19 USD InStock
76462-552 76462-556 CA76462-554 76462-554 CA76462-552 MSPP-SH3102704 CA76462-556
HyClone™ Cell Boost™ 7a and 7b Feed Supplements, HyClone products (Cytiva)
Cell Culture Supplements and Additives
Cell Boost™ 7a and 7b are feed additives especially matched to ActiPro™ production medium. Combining Cell Boost™ 7a and 7b has demonstrated significantly increased productivity when compared with batch culture.
These supplements are used as an essential part of a cell culture feed strategy to enhance cell culture performance and increase product yield. Cell Boost 7a and 7b are intended for use together in defined concentrations. Established protocols facilitate easy and efficient identification of a feeding strategy. Cell Boost 7a contains concentrated amino acids, vitamins, salts, trace elements, and a carbon source. Cell Boost 7b is a balanced formulation of selected amino acids in a concentrated form.
Ordering information: Lead times and minimum order quantities apply. Please contact your local Avantor sales office for more information.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty