Would you like to log in again?
HyClone Cell Boost™ 7b Supplement, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH31120.03 SH31120.02 SH31027.07 SH31027.01 SH31120.02 SH31120.01 SH31120.01 SH31120.03 SH31027.07 SH31027.04 SH31027.01
76462-606EA 520 USD InStock
CA76462-606 76462-606 CA76462-604 CA76534-644 76534-644 76462-556 76462-604 CA76462-602 76462-602 MSPP-SH3102704 CA76462-556
HyClone Cell Boost™ 7b Supplement, HyClone products (Cytiva)
Cell Culture Supplements and Additives
Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.
  • Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pHSupplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
  • Non sterile
Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines. Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty