Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyClone™ CDM4Avian Media, HyClone products (Cytiva)

Supplier: HyClone products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

HyClone™
SH31035.01 SH31035.01
76462-564EA 336.03 USD InStock
76462-564 CA76462-564
HyClone™ CDM4Avian Media, HyClone products (Cytiva)
Cell Culture Media
Designed for vaccine manufacturing using suspension cell culture with avian cell line.

  • Chemically defined (CD) animal-derived component-free (ADCF) formulation
  • Supports high peak cell density as well as enhanced viral infectivity and productivity
  • Economical, operation-reducing single formulation for cell propagation and viral production
  • Designed for small- to large-scale culture applications
  • Off-white powder formulation

HyClone CDM4Avian cell culture medium is developed for manufacturing of viral vaccines in avian cell lines, such as EB66 cells, and for the production of lytic viruses, such as measles, influenza, and Japanese encephalitis (JE); and of non-lytic viruses, such as the modified vaccinia Ankara (MVA) virus.

CDM4Avian is formulated for use in a variety of cell culture platforms, ranging from shaker flasks to WAVE™ and Xcellerex™ bioreactors.

Both powder medium and hydrated medium should be stored at 2 to 8 °C. Product should be stored away from light.

Osmolality: 240 to 280 mOsm/kg H₂O (without NaHCO₃) for powder medium.

Endotoxin: <10 EU/g for powder medium.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty