Would you like to log in again?
HyClone Cell Boost™ 1 Supplement, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH31113.02 SH31113.01 SH31113.02 SH31113.01 SH30584.02 SH30584.02
95026-616EA 312.35 USD InStock
95026-616 76462-574 CA76462-576 76462-576 CA76462-574 CA95026-616
HyClone Cell Boost™ 1 Supplement, HyClone products (Cytiva)
Cell Culture Supplements and Additives
Supplements cell culture with amino acids, vitamins, and glucose and manufactured to meet cGMP manufacturing standards and QC specifications.
  • Designed to support modern cell culture manufacturing processes, Cell Boost 1 is chemically defined (CD), animal-derived component-free (ADCF), and protein-free
  • Does not contain L-glutamine, poloxamer 188, or hydrolysates
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to the dry powder format
Improve productivity and cell growth in fed-batch cultures by supplementing the cell culture medium to boost nutrients depleted during culture. HyClone Cell Boost 1 Supplement provides nutrients such as amino acids, vitamins, and glucose to support fed-batch cultures in research and bioprocessing applications. It is available as a convenient ready-to-use liquid solution or as a dry powder supplement to be hydrated before use.
This cell culture medium supplement has been developed for recombinant protein production with various cell lines including Chinese hamster ovary (CHO) and human embryonic kidney (HEK) 293 cells.
Supplement 100 to 200 ml/L, one or more times, starting during early- to mid-growth phase. This is expected to be day 2 to 4. Start the culture at a reduced volume according to anticipated feed volumes. Continue supplementation daily until the final volume is reached. It is recommended to monitor L- glutamine separately and supplement as needed.
Dry powdered medium milling equivalency study compares pin milling of powdered HyClone cell culture media and supplements at two manufacturing sites. Detailed physical, chemical, and biological data, including cell growth, are provided.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty