Would you like to log in again?
HyClone Cell Boost™ 3 Supplement, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH31115.02 SH31115.01 SH31115.01 SH30825.01 SH31115.02 SH30825.01
76462-582EA 124.66 USD InStock
76462-582 CA76462-584 CA76462-582 CA95026-620 76462-584 95026-620
HyClone Cell Boost™ 3 Supplement, HyClone products (Cytiva)
Cell Culture Supplements and Additives
Supplements cell culture with amino acids, vitamins, glucose, and trace elements and manufactured to meet cGMP manufacturing standards and QC specifications.
  • Designed to support modern manufacturing processes, Cell Boost 3 is chemically defined (CD), animal-derived component-free (ADCF), protein-free, and hydrolysate-free
  • For convenience, this cell culture supplement is available as a ready-to-use liquid solution in addition to a dry powder format
Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 3 Supplement provides amino acids, vitamins, glucose, and trace elements to support fed-batch cultures in research and bioprocessing applications. It is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.
This HyClone cell culture supplement has been developed for recombinant protein production with various cell lines including hybridoma and myeloma cell lines.
Supplement 20 to 200 ml/L one or more times, starting during mid to late growth phase. This is expected to be day 3 to 4. Start the culture at a reduced volume according to anticipated feed volumes. Continue supplementation daily until the final volume is reached. It is recommended to monitor L- glutamine separately and supplement as needed.
Dry powdered medium milling equivalency study compares pin milling of powdered HyClone cell culture media and supplements at two manufacturing sites. Detailed physical, chemical, and biological data, including cell growth, are provided.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty