Would you like to log in again?
Tronex® Spunbond Bouffant Caps
Supplier: Tronex
Tronex®
7124B
76462-936CS 57.38 USD InStock
76462-936
Tronex® Spunbond Bouffant Caps
Disposable Garments
Designed to provide highly comfortable, breathable, and effective protection from fluids and other contaminant, these nonwoven spunbond bouffant caps keep hair securely contained for a clear line of sight.
  • Constructed from a soft, fluid-resistant, non-woven spunbond material for excellent protection
  • Secured with non-binding elastic for comfort
  • Not made with natural rubber latex
Polypropylene spundbond is a fluid-resistant, highly breathable and comfortable non-woven material which provides soft, lightweight protection from fluid exposure.
Packaging: Caps are packaged 100 per box, 5 boxes per case, for a total of 500 caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty