Would you like to log in again?
Tronex® Fluid-Impervious Over-the-Head Gown with Impervious Film and Thumb Hook
Supplier: Tronex
Tronex®
6033C35
76462-942CS 71.58 USD InStock
76462-942
Tronex® Fluid-Impervious Over-the-Head Gown with Impervious Film and Thumb Hook
Disposable Garments
This fluid-impervious over-the-head gown provides excellent protection while retaining comfort and breathability during extended wear.
  • Crafted from a soft, durable, fluid-impervious film material for exceptional protection
  • Full frontal coverage with an open back for ventilation
  • Thumb-hook cuffs for a secure fit and complete arm coverage when worn beneath gloves
  • Apron-style neck opening and perforated back shoulder for easy donning and pull-away removal
  • Waist ties for customized fit
  • Not made with natural rubber latex
Advanced formulation PE material offers enhanced barrier protection for exposure to fluid. Gown features an over-the-head easy breakaway design, with thumb-hooks and waist ties for a secure fit. Packaged in dispenser boxes for efficient stocking and quick access.
Packaging: Gowns are packaged 1 per bag, 15 bags per dispenser box, 5 dispenser boxes per case, for a total of 75 gowns.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty