Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Super Erecta® Stackable Ledge for Wire Shelving, 4" High

Supplier: Intermetro
Super Erecta®
L60N-4K4 L24N-4K4 L48N-4K4 L54N-4K4 L18N-4K4 L42N-4K4 L30N-4K4 L36N-4K4 L14N-4K4 L21N-4K4 L72N-4K4
76463-072EA 77.86 USD InStock
76463-072 76463-062 76463-052 76463-064 76463-054 76463-066 76463-056 76463-068 76463-058 76463-070 76463-060
Super Erecta® Stackable Ledge for Wire Shelving, 4" High
Furniture Shelves
Prevent items from protruding or falling from shelves.

  • Only compatible with wire shelving
  • Ledges may be stacked to form an 8" (203 mm) or higher barrier

Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty