Would you like to log in again?
Super Erecta® Wall Mount Shelving Post with Brackets, Metro™
Supplier: Metro International
Super Erecta®
13PDFK4 33PDFK4 54PDFK4 63PDFK4
76463-074EA 70.64 USD InStock
76463-074 76463-100 76463-102 76463-104
Super Erecta® Wall Mount Shelving Post with Brackets, Metro™
Furniture Shelves
Combine with shelves and shelf supports to create a post-type wall mounted shelving solution.
  • 13" posts are ideal for 1-shelf tier configurations
  • 33" posts support 2-shelf configurations
  • 54" posts can accommodate 3 or more shelving tiers
  • 63" posts are designed to handle 4 or more shelving tiers
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty