Would you like to log in again?
SmartWall® Lid Holder/Drying Shelf, Metro™
Supplier: Metro International
SmartWall®
IWA-11K4
76463-126EA 71.1 USD InStock
76463-126
SmartWall® Lid Holder/Drying Shelf, Metro™
Furniture Shelves
Add a space for drying your lids and dishes to your SmartWall® system.
  • Open-wire design works to prevent wet nesting
  • Each holder is 8.25×13.5×4.5"
  • No tools required to assemble
  • Built to fit ¹/₆ and ¹/₉" size pan lids
Metroseal finish is suitable for wet and damp environments and includes Microban® antimicrobial product protection to inhibit the growth of bacteria, mold, mildew, and fungus that cause odors and product degradation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty