Would you like to log in again?
SmartWall® Wire Grids, Metro™
Supplier: Metro International
SmartWall®
WG3048K4 WG1836K4 WG2430K4 WG3036K4 WG1848K4 WG1830K4 WG2448K4 WG1860K4 WG2436K4
76463-182EA 134.52 USD InStock
76463-182 76463-184 76463-186 76463-176 76463-188 76463-178 76463-190 76463-180 76463-192
SmartWall® Wire Grids, Metro™
Furniture Shelves
Metro® SmartWall® Wire Grids create storage and workstation systems for underutilized wall space.
  • Grids may attach directly to SmartWall® uprights
  • No tools are necessary
  • Grids may not overhang the uprights by more than 6" (152 mm)
  • Grid openings measure approximately 3×3" (76×76 mm)
When SmartWall® uprights are not used, select the appropriate grid bracket kit based on the configuration.
Metroseal Gray epoxy finish comes with Microban® antimicrobial product protection which adds a level of protection against bacteria, mold and mildew that can cause stains, odors, and product degradation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty