Would you like to log in again?
Super Erecta® Industrial Wire Shelves, Metroseal Gray Epoxy
Supplier: Intermetro
Super Erecta®
1842NK4 2160NK4 2124NK4 2136NK4 2424NK4 1836NK4 1448NK4 2472NK4 2454NK4 2130NK4 1860NK4 1830NK4 2460NK4 2448NK4 2148NK4 1472NK4 1436NK4 2442NK4 1430NK4 1424NK4 2154NK4 1824NK4 1854NK4 2430NK4 1442NK4 2172NK4 1848NK4 2436NK4 2142NK4 1872NK4 1460NK4
76463-280EA 209.5 USD InStock
76463-280 76463-282 76463-260 76463-284 76463-240 76463-262 76463-242 76463-264 76463-244 76463-266 76463-224 76463-268 76463-246 76463-226 76463-248 76463-228 76463-270 76463-250 76463-272 76463-230 76463-274 76463-252 76463-254 76463-232 76463-276 76463-256 76463-278 76463-234 76463-258 76463-236 76463-238
Super Erecta® Industrial Wire Shelves, Metroseal Gray Epoxy
Furniture Shelves
Neutral aesthetic for the lab environment, now with corrosion and antimicrobial protection.
  • Shelves can be repositioned at precise 1" (25 mm) increments along the length of the numbered posts
  • Open-wire design minimizes dust accumulation, allows a free circulation of air, and greater visibility of stored items
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial product protection to keep the product 'cleaner between cleanings'.
12-year warranty against rust and corrosion.
Ordering information: Posts and accessories sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty