Neutral aesthetic for the lab environment, now with corrosion and antimicrobial protection.
- Shelves can be repositioned at precise 1" (25 mm) increments along the length of the numbered posts
- Open-wire design minimizes dust accumulation, allows a free circulation of air, and greater visibility of stored items
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial product protection to keep the product 'cleaner between cleanings'.
12-year warranty against rust and corrosion.
Ordering information: Posts and accessories sold separately.