Would you like to log in again?
Starsys® 28" Tall Undercounter Storage Cart
Supplier: Intermetro
Starsys®
SXR28PL1 SXR28NL1 SXR28KL1
76463-286EA 3403.28 USD InStock
76463-286 76463-288 76463-290
Starsys® 28" Tall Undercounter Storage Cart
Carts
Starsys® Undercounter Storage Carts enhance productivity and space efficiency.
  • Durable easy-to- clean, polymer enclosures will not chip, peel, or rust
  • Robust and stable structure accommodates even the most sensitive equipment
  • Security options including non-locking, key locking, and electronic PIN locking with key override
The Starsys® Mobile Undercounter Cart is a polymer based, highly adaptive storage cart, that enhances productivity and space efficiency by empowering the end user with the ability to easily relocate, reposition, and transport stored contents as needed. This ability helps to create a more flexible workspace and to meet your changing workflow demands.
Microban® antimicrobial protection works 24/7 for an added level of protection against bacteria, mold and mildew that can cause stains, odors, and product degradation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty