Would you like to log in again?
Leica Laser Microdissection (LMD) System
Supplier: Leica
Leica®
11600312 11505188 11600288 11505151 11505190 11600312 11505189 11600288 11505151 11505191 11505190 8118616 11505191 11505158 11505215 8118616 11505215 11505158 11505188 11505189
76414-900PK 1571.55 USD InStock
CA76463-318 76414-900 CA76414-778 76414-898 CA76414-776 76414-896 CA76414-774 CA103501-506 76463-320 76463-322 76414-894 CA76463-320 76414-892 76463-324 CA76463-322 CA76414-780 76463-316 CA76463-324 CA76463-316 76463-318
Leica Laser Microdissection (LMD) System
Microtomes and Microtome Assemblies Microtomes
We move the laser, not the sample. And we use gravity for collection. LMD systems provides perfectly cut, contamination-free, analysis-ready dissectates.
  • Unique laser-guidance based dissection method to collect dissectates via gravity – contact-free and contamination-free
  • High-precision optics to steer the laser beam along the desired cut line, leaving clean cuts of contamination-free isolates
  • LMD SmartCut objectives made specifically for the LMD system
  • Save on consumables, as the Leica LMD utilizes standard PCR tubes to capture isolated samples. You will not be locked into expensive consumables with this system
Laser microdissection (LMD, also known as laser capture microdissection or LCM) enables users to isolate specific single cells or entire areas of tissue. Powered by a unique laser design and dynamic software, Leica LMD systems allow users to easily isolate regions of interest (ROI) from entire areas of tissue down to single cells or even subcellular structures such as chromosomes.
LMD is typically used in genomics (DNA), transcriptomics (mRNA, miRNA), proteomics, metabolomics, and even next generation sequencing (NGS). Researchers in neurology, cancer research, plant analysis, forensics or climate research rely on this method. Furthermore, LMD is a perfect tool for live cell culture (LCC), for cloning and re-cultivation, manipulation or downstream analysis.
Certifications: CE and CAS.
Ordering information: For more information on LMD and how we can help with sample preparation of downstream genomic and proteomic analyses, please contact local Avantor representative.
Delivery information: Leica Laser Microdissection (LMD6) System Package includes DM6 upright microscope with motorized Z, CTR6 LED, 7 positon motorized nosepiece for objectives, morotized condenser, S28 condenser head (0.55NA, 28 mm working distance), motorized stage, Basic XYZ-module LX, Smart Move to control motorized stage, objectives (UVI 2.5x/0.07NA Microdissection, UVI 5x/0.12NA Microdissection, HC PL FLUOTAR 10x/0.32NA and HCX PL FL L 20x/0.40NA with correction collar), Patho diffusor plate for 1.25-5x object, Wacom Cintiq 22HD Pen Display, LMD CC7000 camera 11541544, C-mount HC 0.55x, Tube adapter including one documentation port. Purchase of LMD package includes installation and training.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty