Would you like to log in again?
VWR STOPPER CELLULOSE 43-46MM 50MM PK500
Supplier: VWR International
Vwr* Cellulose stopper, Special shape, F TUBE IN D43, Diameter: 43-46mm, made of cellulose and are used for microbiological samples and tissue cultures in test tubes or Erlenmeyer flasks, Air permeable, Autoclavable up to 200 deg C, Filters bacteria, Dimension: 50x41.5x50mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR STOPPER CELLULOSE 43-46MM 50MM PK500 76463-378 Pack of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty