Would you like to log in again?
TIP PIPET FILTERED BLACK 300UL CS5760
Supplier: AZER SCIENTIFIC INC
Pipet tip, Robotic filter, for automation, Hamilton compatible, conductive 30ul filter tips, Color: Black, Tip Shape: Round, Polypropylene, Nonpyrogenic, Non-sterile, Liquid Level Sensing Tips, 96/rack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP PIPET FILTERED BLACK 300UL CS5760 76463-402 ES81290-0300FR Case of 5,760 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty