Would you like to log in again?
VWR® Disposable Scrub Pants
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
7389 7284W 7289 7285 7281W 72899 73899 7289 7284 7283 7282W 7382 7288 7386 7380 7289W 7381 7282 7388 7281 7286 7287 7384 7283W 7288 7383 7280 7286W 7387 7288W 7385 7280W 7287W 7285W
76463-418CS 261.83 USD InStock
76463-418 76006-542 76006-544 75832-236 76006-540 75832-238 75832-242 76006-538 75832-244 76006-556 76006-534 75832-240 76006-536 76463-420 76463-422 76463-424 76463-406 76463-408 76006-552 76006-532 76006-554 75832-246 75832-248 76006-550 75832-232 75832-234 76006-546 75832-250 76006-548 75832-252 76463-410 76463-412 76463-414 76463-416
VWR® Disposable Scrub Pants
Disposable Garments
VWR® Disposable Scrub Pants are an ideal option for single-use needs.
  • Elastic Waist
  • Single Use
  • Hip Pockets Available
  • USA Made
  • Perfect with VWR® Disposable Scrub Shirts
These garments feature high-quality sewn-seam construction and are made from SMS nonwoven fabric for comfort.
The versatile pants help meet PPE requirements and provide comfort at the same time.
Packaging: These blue scrubs pants are packaged 30 per case within a double bag-lined carton.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty