Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYP1A1 Rabbit Polyclonal Antibody (DyL550)

Supplier: Boster
A00471-DYL550
76463-732EA 378.14 USD InStock
76463-732
Anti-CYP1A1 Rabbit Polyclonal Antibody (DyL550)
Antibodies
Rabbit IgG Polyclonal Anti-Human CYP1A1 Antibody DyLight®, 550 Conjugated, Flow Validated.

Type: Primary
Antigen: CYP1A1
Clonality: Polyclonal
Clone:
Conjugation: DyL550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty