Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cyp1a2 Rabbit Polyclonal Antibody

Supplier: Boster
A00598
76463-826EA 360.95 USD InStock
76463-826
Anti-Cyp1a2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Cytochrome P450 1A2/Cyp1a2 detection. Tested with WB, FCM, Direct ELISA in Mouse,Rat.

Type: Primary
Antigen: Cyp1a2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse,Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty