Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NRXN1 Rabbit Polyclonal Antibody (DyL488)

Supplier: Boster
A01490-DYL488
76464-412EA 378.14 USD InStock
76464-412
Anti-NRXN1 Rabbit Polyclonal Antibody (DyL488)
Antibodies
Rabbit IgG Polyclonal Anti-Human Neurexin 1 Antibody DyLight®, 488 Conjugated, Flow Validated.

Type: Primary
Antigen: NRXN1
Clonality: Polyclonal
Clone:
Conjugation: DyL488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty