Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Wfdc2 Rabbit Polyclonal Antibody (Biotin)

Supplier: Boster
A02685-BIOTIN
76464-974EA 360.95 USD InStock
76464-974
Anti-Wfdc2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Rabbit IgG polyclonal antibody for HE4 detection. Tested with ELISA(Det) in Rat.

Type: Primary
Antigen: Wfdc2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty