Would you like to log in again?
Anti-Lymphotactin Polyclonal Antibody
Supplier: Boster
A04548
76465-368EA 360.95 USD InStock
76465-368
Anti-Lymphotactin Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Lymphotactin detection. Tested with WB, Direct ELISA in Mouse;Rat.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty