Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SETDB2 Rabbit Polyclonal Antibody

Supplier: Boster
A08290-2
76465-750EA 360.95 USD InStock
76465-750
Anti-SETDB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for CLLD8/SETDB2 detection. Tested with WB, FCM in Human,Mouse,Rat.

Type: Primary
Antigen: SETDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty