Would you like to log in again?
Cell Counting Kit-8 (CCK-8)
Supplier: Boster
AR1160
76466-042EA 91.67 USD InStock
76466-042
Cell Counting Kit-8 (CCK-8)
Assays Cellular Assays
Cell counting kit-8 (CCK-8) allows very convenient assays by utilizing highly water-soluble tetrazolium salt. WST-8 [2- (2- methoxy-4-nitrophenyl)-3- (4-nitrophenyl)-5- (2, 4-disulfophenyl)-2H- tetrazolium, monosodium salt]* produces a water-soluble formazan dye upon reduction in the presence of an electron carrier. Cell Counting Kit-8 is a one-bottle solution; no premixing of components is required.
  • One bottle, ready to use solution
  • No organic solvents or isotopes required
  • No harvesting, no washing and no solubilization steps
  • No toxicity to cell
  • More sensitive than MTT, XTT, MTS or WST-1
Cell Counting Kit-8, being nonradioactive, allows sensitive colorimetric assays for the determination of the number of viable cells in cell proliferation cytotoxicity assays. WST-8 is reduced by dehydrogenases in cells to give an orange colored product (formazan), which is soluble in the tissue culture medium. The amount of the formazan dye generated by the activity of dehydrogenases in cells is directly proportional to the number of living cells.
The cell proliferation assay using CCK-8 correlates well with the [(3)H] thymidine incorporation assay. And the CCK-8 assay can also be substituted for the [(3)H]thymidine incorporation assay. The detection sensitivity of CCK-8 is higher than other tetrazolium salts such as MTT, XTT, MTS or WST-1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty