Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GL50803_14482 Rabbit Polyclonal Antibody

Supplier: Boster
DZ00135
76466-120EA 401.06 USD InStock
76466-120
Anti-GL50803_14482 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for eIF4E detection.

Type: Primary
Antigen: GL50803_14482
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Giardia lamblia

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty