Would you like to log in again?
Mouse K-Cadherin-6 Cdh6 PicoKine; ELISA Kit, Boster
Supplier: Boster
PicoKine
EK2097
76467-060EA 422.82 USD InStock
76467-060
Mouse K-Cadherin-6 Cdh6 PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for quantitative detection of mouse Cadherin-6. 96 wells/kit, with removable strips.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty