Would you like to log in again?
Human LAIR1 PicoKine; ELISA Kit, Boster
Supplier: Boster
PicoKine
EK2111
76467-086EA 422.82 USD InStock
76467-086
Human LAIR1 PicoKine; ELISA Kit, Boster
Assays ELISAs
For quantitative detection of human LAIR1 in cell culture supernatants, serum and plasma (heparin, EDTA, citrate).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty