Would you like to log in again?
Mouse Angiopoietin-1 ELISA Kit EZ-Set (DIY Anti-body Pairs), Boster
Supplier: Boster
EZ1296
76467-256EA 292.2 USD InStock
76467-256
Mouse Angiopoietin-1 ELISA Kit EZ-Set (DIY Anti-body Pairs), Boster
Assays ELISAs
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Angiopoietin-1 in Mouse samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty