Would you like to log in again?
Mouse OPG (TNFRSF11B) / Osteoprotegerin PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0481
76467-344EA 571.78 USD InStock
76467-344
Mouse OPG (TNFRSF11B) / Osteoprotegerin PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Mouse Osteoprotegerin/TNFRSF11B with <5 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum and plasma (EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Osteoprotegerin/TNFRSF11B tissue specificity: Highly expressed in liver, lung, stomach, intestines and calvaria. Highly expressed in decidua and placenta, and in embryo.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty