Would you like to log in again?
Mouse Adiponectin PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0596
76467-378EA 571.78 USD InStock
76467-378
Mouse Adiponectin PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Quick Picokine ELISA kits, assay takes less than 1.5 hours. Detect Mouse Adiponectin with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum, plasma (heparin, EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and quick experiment set up.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty