Would you like to log in again?
Human CD163 / M130 PicoKine; Quick ELISA Kit, Boster Technology
Supplier: Boster
PicoKine
FEK0715
76467-386EA 571.78 USD InStock
76467-386
Human CD163 / M130 PicoKine; Quick ELISA Kit, Boster Technology
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours.
Detect Human CD163 with <150 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum and plasma (heparin). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. CD163 tissue specificity: Expressed in monocytes and mature macrophages such as Kupffer cells in the liver, red pulp macrophages in the spleen, cortical macrophages in the thymus, resident bone marrow macrophages and meningeal macrophages of the central nervous system. Expressed also in blood. Isoform 1 is the lowest abundant in the blood. Isoform 2 is the lowest abundant in the liver and the spleen. Isoform 3 is the predominant isoform The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detected in the blood.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty