Would you like to log in again?
Human PLAT/TPA PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK0897
76467-400EA 571.78 USD InStock
76467-400
Human PLAT/TPA PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Human Tpa/PLAT with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum, plasma (heparin, EDTA, citrate) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tpa/PLAT tissue specificity: Synthesized in numerous tissues (including tumors) and secreted into most extracellular body fluids, such as plasma, uterine fluid, saliva, gingival crevicular fluid, tears, seminal fluid, and milk.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty