Would you like to log in again?
Human Ficolin-3 PicoKine; Quick ELISA Kit, Boster
Supplier: Boster
PicoKine
FEK1259
76467-430EA 571.78 USD InStock
76467-430
Human Ficolin-3 PicoKine; Quick ELISA Kit, Boster
Assays ELISAs
The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Human Ficolin-3/FCN3 with <20 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum and plasma (heparin). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Ficolin-3/FCN3 tissue specificity: Liver and lung. In liver it is produced by bile duct epithelial cells and hepatocytes. In lung it is produced by both ciliated bronchial epithelial cells and type II alveolar epithelial cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty